missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorothieno[2,3-d]pyrimidine, Tech., Thermo Scientific™

1065.00 NOK - 6175.00 NOK

Chemical Identifiers

CAS 14080-59-2
Molecular Formula C6H3ClN2S
Molecular Weight (g/mol) 170.61
MDL Number MFCD01312373
InChI Key NZCRUBBNZGVREM-UHFFFAOYSA-N
PubChem CID 736618
IUPAC Name 4-chlorothieno[2,3-d]pyrimidine
SMILES ClC1=C2C=CSC2=NC=N1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10064662
SDS View Documents
Thermo Scientific Maybridge
GK02802DA
1 g
1065.00 NOK
1g
Please to purchase this item. Need a web account? Register with us today!
10351735
SDS View Documents
Thermo Scientific Maybridge
GK02802DE
5 g
2877.90 NOK
5g
Please to purchase this item. Need a web account? Register with us today!
10046252
SDS View Documents
Thermo Scientific Maybridge
GK02802EA
10 g
6175.00 NOK
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Chlorothieno[2,3-d]pyrimidine, Tech., Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.