missing translation for 'onlineSavingsMsg'
Learn More
4-Chloro-3-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™ Product Code.: 11376979

4-Chloro-3-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™

Product Code. 11376979
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11376979

Brand: Thermo Scientific Alfa Aesar H26834.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261763-23-9
Molecular Formula C8H5BrClF3
Molecular Weight (g/mol) 273.477
MDL Number MFCD01631540
InChI Key LZLIPLUATRVXSB-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl benzyl bromide, 4-bromomethyl-1-chloro-2-trifluoromethyl benzene, 3-trifluoromethyl-4-chlorobenzyl bromide, 4-bromomethyl-1-chloro-2-trifluoromethyl-benzene, benzene, 4-bromomethyl-1-chloro-2-trifluoromethyl, acmc-20amlg, pubchem3030, 3-trifluoromethyl-4-chlorobenzylbromide, 3-trifluromethyl-4-chlorobenzyl bromide, 4-chloro-3-trifluoromethylbenzyl bromideShow MoreShow Less
PubChem CID 2778114
IUPAC Name 4-(bromomethyl)-1-chloro-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1CBr)C(F)(F)F)Cl

Specifications

Melting Point 36°C to 40°C
Quantity 1 g
UN Number UN3261
Formula Weight 273.48
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)benzyl bromide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10150751 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot NumberB25T066 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10158118 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10137801 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10142842 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Chloro-3-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.