missing translation for 'onlineSavingsMsg'
Learn More
4-Chloro-2-fluorophenylacetic acid, 97%, Thermo Scientific™ Product Code.: 11495900

4-Chloro-2-fluorophenylacetic acid, 97%, Thermo Scientific™

Product Code. 11495900
5 g, 5g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11495900

Brand: Thermo Scientific Alfa Aesar H26512.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Chloro-2-fluorophenylacetic acid is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 194240-75-0
Molecular Formula C8H6ClFO2
Molecular Weight (g/mol) 188.58
MDL Number MFCD03095402
InChI Key WKAZXGAJEGPUAY-UHFFFAOYSA-N
Synonym 4-chloro-2-fluorophenylacetic acid, 2-4-chloro-2-fluorophenyl acetic acid, 4-chloro-2-fluorophenyl acetic acid, 4-chloro-2-fluorophenylaceticacid, rarechem al bo 0928, pubchem4292, acmc-20a3w8, ksc541i7t, 4-chlor-2-flour phenyactic acid, 4-chloro-2-fluoro-phenyl acetic acid
PubChem CID 2757563
IUPAC Name 2-(4-chloro-2-fluorophenyl)acetic acid
SMILES OC(=O)CC1=CC=C(Cl)C=C1F

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 188.59
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-fluorophenylacetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Chloro-2-fluorophenylacetic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.