missing translation for 'onlineSavingsMsg'
Learn More
(4-Bromophenoxy)-tert-butyldimethylsilane, 97% Product Code.: 10031544

(4-Bromophenoxy)-tert-butyldimethylsilane, 97%

Product Code. p-4090024 Shop All Thermo Scientific Chemicals Products
5 mL, Glass bottle, 5mL
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
This item is not returnable. View return policy

Product Code. 10031544

Brand: Thermo Scientific Acros 427090050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 67963-68-2 | C12H19BrOSi | 287.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67963-68-2
Molecular Formula C12H19BrOSi
Molecular Weight (g/mol) 287.27
MDL Number MFCD00239382
InChI Key DLGZGLKSNRKLSM-UHFFFAOYSA-N
Synonym 4-bromophenoxy tert-butyl dimethylsilane, 4-bromophenoxy-tert-butyldimethylsilane, p-bromophenoxy t-butyl dimethylsilane, 4-bromophenoxy-tert-butyl-dimethylsilane, 1-bromo-4-tert-butyldimethylsiloxy benzene, 4-bromophenol tert-butyldimethylsilyl ether, silane, 4-bromophenoxy 1,1-dimethylethyl dimethyl, 1-bromo-4-1,1-dimethylethyl dimethylsilyl oxy benzene, acmc-1b8oo
PubChem CID 4227167
IUPAC Name (4-bromophenoxy)-tert-butyl-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)OC1=CC=C(C=C1)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 1.1740g/mL
Boiling Point 137°C (25.0 mmHg)
Flash Point 12°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4OSi(CH3)2C(CH3)3
Quantity 5 mL
Specific Gravity 1.174
Formula Weight 287.27
Percent Purity 97%
Chemical Name or Material (4-Bromophenoxy)-tert-butyldimethylsilane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Product Code
Lot Number334153 Certificate TypeCertificate of Analysis Date05/01/2023 Product Code427090250,427090050
Lot Number334153 Certificate TypeCertificate of Analysis Date12/12/2022 Product Code427090250,427090050
compliance-icons compliance-icons
Product Identifier
  • (4-Bromophenoxy)-tert-butyldimethylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(4-Bromophenoxy)-tert-butyldimethylsilane, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.