Learn More
4-Bromo-2-fluoro-1-(trifluoromethoxy)benzene, 97%, Thermo Scientific™
Product Code. 11425880
Click to view available options
Quantity:
1 g
10 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 105529-58-6
Molecular Formula C7H3BrF4O
Molecular Weight (g/mol) 258.998
MDL Number MFCD01861132
InChI Key SBSFDYRKNUCGBZ-UHFFFAOYSA-N
Synonym 4-bromo-2-fluoro-1-trifluoromethoxy benzene, 1-bromo-3-fluoro-4-trifluoromethoxy benzene, 1-bromo-3-fluoro-4-trifluoromethoxybenzene, 2-fluoro-4-bromotrifluoromethoxybenzene, 4-bromo-2-fluorotrifluoromethoxybenzene, benzene, 4-bromo-2-fluoro-1-trifluoromethoxy, 3-fluoro-4-trifluoromethoxy bromobenzene, 3-bromo-2-fluorotrifluoromethoxybenzene, 4-bromo-2-fluorophenyl trifluoromethyl ether, 4-bromo-alpha,alpha,alpha,2-tetrafluoroanisole
PubChem CID 2782217
IUPAC Name 4-bromo-2-fluoro-1-(trifluoromethoxy)benzene
SMILES C1=CC(=C(C=C1Br)F)OC(F)(F)F
Specifications
Refractive Index 1.447
Quantity 1 g
Formula Weight 259
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-fluoro-1-(trifluoromethoxy)benzene