missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,5-difluorobenzenesulfonyl chloride, 98%, Thermo Scientific™

Product Code. p-3790884 Shop All Thermo Scientific Chemicals Products
1 g, Glass bottle, 1g
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 207974-14-9
Molecular Formula C6H2BrClF2O2S
Molecular Weight (g/mol) 291.49
MDL Number MFCD00042181
InChI Key SBMKFWMFNIEPDN-UHFFFAOYSA-N
Synonym 4-bromo-2,5-difluorobenzene-1-sulfonylchloride, 4-bromo-2,5-difluorobenzenesulphonyl chloride, 4-bromo-2,5-difluorobenzene-1-sulfonyl chloride, 4-bromo-2,5-difluorophenyl chlorosulfone, 4-bromo-2,5-difluorobenzenesulphonylchloride, benzenesulfonyl chloride, 4-bromo-2,5-difluoro, 4-bromo-3,6-difluoro-benzenesulfonylchloride, pubchem5058, acmc-209feg, 4-bromo-2,5-difluorobenzenesulfonylchloride
PubChem CID 574590
IUPAC Name 4-bromo-2,5-difluorobenzenesulfonyl chloride
SMILES FC1=CC(=C(F)C=C1Br)S(Cl)(=O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 37°C to 39°C
Color Orange or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5 % min. (GC)
Packaging Glass bottle
Linear Formula F2C6H2(Br)SO2Cl
Quantity 1 g
Formula Weight 291.49
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Bromo-2, 5-difluorobenzenesulfonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-2,5-difluorobenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Bromo-2,5-difluorobenzenesulfonyl chloride, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.