missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzoic acid (PABA), 98.5 to 101.5% (Dry Basis), Thermo Scientific™

1076.00 NOK - 8448.00 NOK

Chemical Identifiers

CAS 150-13-0
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00007894
InChI Key ALYNCZNDIQEVRV-UHFFFAOYSA-N
Synonym p-aminobenzoic acid, paba, para-aminobenzoic acid, vitamin bx, sunbrella, p-carboxyaniline, 4-carboxyaniline, hachemina, paraminol, benzoic acid, 4-amino
PubChem CID 978
ChEBI CHEBI:30753
IUPAC Name 4-aminobenzoic acid
SMILES NC1=CC=C(C=C1)C(O)=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15895078
SDS View Documents
Thermo Scientific
J11055.36
500 g Plastic bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15895248
SDS View Documents
Thermo Scientific
J11055.A1
1 kg Plastic bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15805258
SDS View Documents
Thermo Scientific
J11055.A7
5 kg Plastic bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 205-753-0

missing translation for 'rtecsNumber' : DG1400000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Aminobenzoic acid (PABA), 98.5 to 101.5% (Dry Basis), Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.