missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-6-hydroxy-1H-pyrazolo[3,4-d]pyrimidine, 90+%, Thermo Scientific™

Product Code. 11332266
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5472-41-3
Molecular Formula C5H5N5O
Molecular Weight (g/mol) 151.129
MDL Number MFCD00005689
InChI Key KTQYLKORCCNJTQ-UHFFFAOYSA-N
Synonym 4-amino-6-hydroxypyrazolo 3,4-d pyrimidine, 4-amino-1h-pyrazolo 3,4-d pyrimidin-6-ol, 4-aminopyrazolo 3,4-d pyrimidin-6-ol, 4-amino-2h-pyrazolo 3,4-d pyrimidin-6-ol, 4-amino-1h-pyrazolo 3,4-d pyrimidin-6 7h-one, 4-aminopyrazolo 5,4-d pyrimidin-6-ol, acmc-1akdw, 4-amino-6-hydroxypyrazolo-3,4-d pyrimidine, 4-amino-1h-pyrazolo 4,3-e pyrimidin-6-ol, 4-amino-7-hydroxypyrazolo 3,4-d pyrimidine
PubChem CID 79623
IUPAC Name 4-amino-1,2-dihydropyrazolo[3,4-d]pyrimidin-6-one
SMILES C1=C2C(=NC(=O)N=C2NN1)N

Specifications

Melting Point >300°C
Odor Odorless
Quantity 1 g
Beilstein 152192
Formula Weight 151.13
Percent Purity ≥90%
Chemical Name or Material 4-Amino-6-hydroxy-1H-pyrazolo[3,4-d]pyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'