missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-2-chloro-3-iodopyridine, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifica
Specifica
Melting Point | 116°C to 117°C |
Color | Orange-Pink to White |
Quantity | 1 g |
Sensitivity | Light sensitive |
Formula Weight | 254.46 |
Percent Purity | 95% |
Physical Form | Solid |
Chemical Name or Material | 4-Amino-2-chloro-3-iodopyridine |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10177586 | Certificate of Analysis | 08/12/2021 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
4-Amino-2-chloro-3-iodopyridine, 95%, Thermo Scientific™ >
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto