missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Dihydro-1,3-benzodioxine-6-amine, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Quantity | 5 g |
IUPAC Name | 4H-1,3-benzodioxin-6-amine |
Formula Weight | 151.16 |
Percent Purity | 97% |
Chemical Name or Material | 4, 5-Dihydro-1, 3-benzodioxine-6-amine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4,5-Dihydro-1,3-benzodioxine-6-amine, ≥97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction