Learn More
4,5-Dibromo-2-phenyl-2,3-dihydropyridazin-3-one, 97%, Thermo Scientific™ Product Code.: 10714763
4,5-Dibromo-2-phenyl-2,3-dihydropyridazin-3-one, 97%, Thermo Scientific™
Product Code. 10714763
10 g, Amber Glass Bottle, 10g
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
Product Code. 10714763
Brand: Thermo Scientific Maybridge BTB07379EA
Request Bulk or Custom Format
Chemical Identifiers
CAS 14305-08-9
Molecular Formula C10H6Br2N2O
Molecular Weight (g/mol) 329.979
MDL Number MFCD00052907
InChI Key NQJXEMRMTOSSBQ-UHFFFAOYSA-N
Synonym 4,5-dibromo-2-phenylpyridazin-3 2h-one, 4,5-dibromo-2-phenyl-2,3-dihydropyridazin-3-one, 4,5-dibromo-2-phenyl-3 2h-pyridazinone, 3 2h-pyridazinone, 4,5-dibromo-2-phenyl, 4,5-dibromo-2-phenyl-3-pyridazinone, 4,5-dibromo-2-phenyl-pyridazin-3-one, 2-phenyl-4,5-dibromo-3 2h-pyridazinone, 3 2h-pyridazinone,4,5-dibromo-2-phenyl, 4,5-bis bromanyl-2-phenyl-pyridazin-3-one, 4,5-dibromo-2-phenyl-2-hydropyridazin-3-one
PubChem CID 203396
IUPAC Name 4,5-dibromo-2-phenylpyridazin-3-one
SMILES C1=CC=C(C=C1)N2C(=O)C(=C(C=N2)Br)Br
Specifications
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 329.98
Percent Purity 97%
Chemical Name or Material 4, 5-Dibromo-2-phenyl-2, 3-dihydropyridazin-3-one
Product Content Correction
Product Title
4,5-Dibromo-2-phenyl-2,3-dihydropyridazin-3-one, 97%, Thermo Scientific™ >