Learn More
4,5,6,7-Tetrahydrobenzo[b]thiophene-3-carboxylic acid, 97%, Thermo Scientific™
1806.51 NOK - 7468.00 NOK
Chemical Identifiers
CAS | 19156-54-8 |
---|---|
Molecular Formula | C9H10O2S |
Molecular Weight (g/mol) | 182.237 |
MDL Number | MFCD00652575 |
InChI Key | TUZZQEHPGHKGRJ-UHFFFAOYSA-N |
Synonym | 4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylic acid, 4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylic acid, 4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylicacid, benzo b thiophene-3-carboxylic acid, 4,5,6,7-tetrahydro, benzo b thiophene-3-carboxylicacid, 4,5,6,7-tetrahydro, 4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylic a, 4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylicacid |
PubChem CID | 767738 |
IUPAC Name | 4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylic acid |
SMILES | C1CCC2=C(C1)C(=CS2)C(=O)O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
19156-54-8 | |
182.237 | |
TUZZQEHPGHKGRJ-UHFFFAOYSA-N | |
767738 | |
C1CCC2=C(C1)C(=CS2)C(=O)O |
C9H10O2S | |
MFCD00652575 | |
4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylic acid, 4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylic acid, 4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylicacid, benzo b thiophene-3-carboxylic acid, 4,5,6,7-tetrahydro, benzo b thiophene-3-carboxylicacid, 4,5,6,7-tetrahydro, 4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylic a, 4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylicacid | |
4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylic acid |
Specifications
19156-54-8 | |
MFCD00652575 | |
4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylic acid, 4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylic acid, 4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylicacid, benzo b thiophene-3-carboxylic acid, 4,5,6,7-tetrahydro, benzo b thiophene-3-carboxylicacid, 4,5,6,7-tetrahydro, 4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylic a, 4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylicacid | |
C1CCC2=C(C1)C(=CS2)C(=O)O | |
182.237 | |
182.24 | |
4,5,6,7-Tetrahydrobenzo[b]thiophene-3-carboxylic acid |
C9H10O2S | |
1367376 | |
TUZZQEHPGHKGRJ-UHFFFAOYSA-N | |
4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylic acid | |
767738 | |
97% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.