missing translation for 'onlineSavingsMsg'
Learn More
4-(4-Methoxyphenyl)-2-butanone, 99% Product Code.: 11434483

4-(4-Methoxyphenyl)-2-butanone, 99%

Product Code. 11434483
50 g, 50g
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11434483

Brand: Thermo Scientific Alfa Aesar A11011.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 104-20-1 | C11H14O2 | 178.23 g/mol

4-(4-Methoxyphenyl)-2-butanone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Methoxyphenyl)-2-butanone is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-20-1
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00008791
InChI Key PCBSXBYCASFXTM-UHFFFAOYSA-N
Synonym 4-4-methoxyphenyl-2-butanone, anisylacetone, 4-4-methoxyphenyl butan-2-one, 4-methoxybenzylacetone, raspberry ketone methyl ether, 2-butanone, 4-4-methoxyphenyl, anisylacetone, p, 4-p-methoxyphenyl-2-butanone, methyl oxanone, 1-4-methoxyphenyl-3-butanone
PubChem CID 61007
IUPAC Name 4-(4-methoxyphenyl)butan-2-one
SMILES COC1=CC=C(CCC(C)=O)C=C1

Specifications

Melting Point 7°C to 9°C
Density 1.046
Boiling Point 147°C to 148°C (8 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.52
Quantity 50 g
Beilstein 1869592
Solubility Information Not miscible in water.
Formula Weight 178.23
Percent Purity 99%
Chemical Name or Material 4- (4-Methoxyphenyl)-2-butanone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number10139481 Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA11011.30,A11011.09,A11011.18
Lot Number10117882 Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA11011.30,A11011.09,A11011.18
Lot Number10165584 Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA11011.30,A11011.09,A11011.18
Lot NumberB8460A Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA11011.30,A11011.09,A11011.18
Lot NumberB8460B Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA11011.30,A11011.09,A11011.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(4-Methoxyphenyl)-2-butanone, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.