Learn More
Memmert™ 4 - 20 mA Current Loop Interface O2 Controller
4-20mA current loop interface for O2 control option
2845.00 NOK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
17109974
|
Memmert™
ICO150V1 |
ICO150 |
N/A
|
N/A | ||||||
17140005
|
Memmert™
ICO240V1 |
ICO240 |
2845.00 NOK
Each |
Estimated Shipment: 20-08-2025
Log in to see stock. |
||||||
17170035
|
Memmert™
ICO50V1 |
ICO50 |
N/A
|
N/A | ||||||
17179934
|
Memmert™
ICO105V1 |
ICO105 |
2845.00 NOK
Each |
Estimated Shipment: 20-08-2025
Log in to see stock. |
||||||
Description
- Requires option T6
- 0 to 25 % O2 = 4 - 20 mA
Specifications
Requires option T6 |
Product Title
Memmert™ 4 - 20 mA Current Loop Interface O2 Controller