missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 4-(2-Pyridylazo)resorcinol monosodium salt monohydrate, 99+%

CAS: 16593-81-0 | C11H9N3O2 | 215.21 g/mol

1387.36 NOK - 12403.92 NOK

Specifications

Chemical Name or Material 4-(2-Pyridylazo)resorcinol monosodium salt monohydrate
CAS 16593-81-0, 1141-59-9
CAS Min % 99.0
CAS Max % 100.0
Color Brown to Brown-Orange
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10592771
View Documents
Thermo Scientific Chemicals
176320050
5 g Glass bottle
1387.36 NOK
5g
Please to purchase this item. Need a web account? Register with us today!
10144050
View Documents
Thermo Scientific Chemicals
176320250
25 g Glass bottle
5055.63 NOK
25g
Please to purchase this item. Need a web account? Register with us today!
10239153
View Documents
Thermo Scientific Chemicals
176321000
100 g Glass bottle
12403.92 NOK
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

4-(2-Pyridylazo)resorcinol monosodium salt monohydrate
99.0
Brown to Brown-Orange
99+%
C11H9N3O2
22, I, 694
4-2-pyridylazo resorcinol monosodium saltmonohydrate
VLCAILLZPUINNF-UKTHLTGXSA-N
3-hydroxy-4-(pyridin-2-ylhydrazinylidene)cyclohexa-2,5-dien-1-one;sodium;hydrate
132988475
99+%
16593-81-0, 1141-59-9
100.0
Authentic
Glass bottle
MFCD00150712,MFCD00150712,MFCD00150712,MFCD00006256
15, 7125
(0.1% in water) Clear
OC1=CC(=O)C=C\C1=N/NC1=CC=CC=N1
215.21
255.21
Crystalline Powder
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals 4-(2-Pyridylazo)resorcinol monosodium salt monohydrate, 99+%

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.