missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[4-(1H-Pyrazol-1-yl)phenyl]methanol, 97%, Thermo Scientific™
Click to view available options
Amber Glass Bottle; 1g
Specifications
Specifications
CAS | 143426-49-7 |
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Molecular Formula | C10H10N2O |
MDL Number | MFCD02682057 |
Quantity | 1 g |
Synonym | 4-1h-pyrazol-1-yl phenyl methanol, 4-pyrazol-1-yl-phenyl methanol, 4-pyrazol-1-ylphenyl methanol, 4-1-pyrazolyl phenyl methanol, 4-1-pyrazolyl benzylalcohol, 4-pyrazol-1-yl phenyl methanol, 4-1h-pyrazol-1-yl benzyl alcohol, benzenemethanol, 4-1h-pyrazol-1-yl, 4-pyrazolylphenyl methan-1-ol |
InChI Key | MMGMLIHSHFKRDK-UHFFFAOYSA-N |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 10
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
[4-(1H-Pyrazol-1-yl)phenyl]methanol, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction