Learn More
3M™ Versaflo™ TR-Series PAPR Accessory, Spark Arrestor
When installed inside the filter cover of the TR-300 PAPR, the TR-362 spark arrestor helps protect the HE filter and PAPR unit from damage due to sparks or other hot materials.
Brand:  3M™ 7000103970
105.19 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15709897
  • 181.00 NOK / Each
Estimated Shipment: 24-07-2025
Description
  • 3M Catalog# TR-362
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Reusable
Protects The he Filter and Powered Air Purifying Respirator Unit From Damage Due to Sparks or Other Hot Materials
TR-362
Spark Arrestor
Versaflo TR-Series PAPR System
2/Cs.
Videos
  • 3M - Powered and supplied air products
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      3M Powered and supplied air products
    Certificates
    Promotions
    Promotions
    Product Content Correction
    Product Title
    3M™ Versaflo™ TR-Series PAPR Accessory, Spark Arrestor > Spark Arrestor/Prefilter