Learn More
3M™ Qualitative Fit-Test Kit Accessories
For use with 3M™ Qualitative Fit Test Apparatus
Brand:  3M™ 7000001945
1516.70 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15135157
  • 2575.00 NOK / Each
Only null left
Estimated Shipment: 18-07-2025
Description
  • Test kits and accessories designed to help your organization meet OSHA fit testing requirements for respirators
  • The products can be used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate prefilter
  • 3M Catalog# 601, FT-13, FT-14, FT-15
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fit Test Adapter
Quantitative Fit Testing
601
OSHA
Qualitative Fit Test Apparatus FT10 and FT30
Videos
  • Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      3M Fit test kit training video
    Certificates
    Promotions
    Promotions
    Product Content Correction
    Product Title
    3M™ Qualitative Fit-Test Kit Accessories > Adapter