Learn More
Corning™ 384-Well, Cell Culture-Treated, Flat-Bottom, Low Flange, Bar-Coded Microplate
Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid
Brand: Corning™ 3764BC
This item is not returnable.
View return policy
Description
- Opaque walls to prevent well-to-well crosstalk
- Optically clear flat well bottom permits direct microscopic viewing
- Can be used for both top and bottom reading instruments
- Flat bottoms with 112μL total volume
- Recommended working volumes of 20 to 80μL
- TC treated for optimal cell attachment
- Sterilized by gamma radiation and certified nonpyrogenic
- Individual alphanumeric codes for well identification
- Bulk packed 20 per bag
Specifications
Standard | |
16 x 24 | |
Polystyrene | |
20 to 80 μL | |
Cell Culture | |
20/Pk. |
With Lid | |
Black, Clear Bottom | |
384 | |
Sterile | |
Square, Flat Bottom (F-well) |
Product Content Correction
Product Title
Corning™ 384-Well, Cell Culture-Treated, Flat-Bottom, Low Flange, Bar-Coded Microplate > Blac; TC treated; 20 to 80uL working volume