Learn More
Corning™ 384 Well Flat Clear Bottom Polystyrene NBS™ Microplates, low flange, no lid, nonsterile
Ideal for reducing protein and nucleic acid binding at low concentrations, and increasing assay signal to noise. Corning™ 384 Well Flat Clear Bottom Polystyrene NBS™ Microplates feature black wall plates that have low background fluorescence, minimal light scatter, and reduced crosstalk.
Brand: Corning™ 3766
17690.50 NOK valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
- Black wall plates have low background fluorescence, minimal light scatter, and reduced crosstalk
- Nonbinding (NBS™) surface is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimized molecular interactions
- Ideal for reducing protein and nucleic acid binding at low concentrations, and increasing assay signal to noise
- Suited for fluorescent assays using either top or bottom detection plate readers
- Recommended working volume of up to 20 to 80μL
- Conforms to standard microplate footprint and dimensions
Specifications
Standard | |
Black | |
384 | |
Non-sterile | |
Square, Flat Bottom (F-well) |
16 x 24 | |
Polystyrene | |
20 to 80 μL | |
Non-binding | |
25 |
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
New Offer Save Up to 92%