Learn More
Sampling Systems™ 316 Stainless Steel Storage Containers without Toggle Clamps
Multi-purpose containers with light duty construction for a wide range of storage uses in the laboratory.
Brand: Sampling Systems™ A466-20
This item is not returnable.
View return policy
Description
- Crevice-free interior
- Two sturdy handles
- Close fitting recessed lid with handle
- Flat bottom with chime ring to give added protection
- Toggle clamps and seals are optional extras
Specifications
Yes | |
Recessed Lid | |
Stainless Steel | |
265 mm |
Storage container | |
20 L | |
390 mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Sampling Systems™ 316 Stainless Steel Storage Containers without Toggle Clamps > Capacity: 20L