Learn More
RETSCH 305 dia. x 40 mmH Stainless Steel Test Sieve with Square Holes
Brand:  RETSCH 60.138.050000
 View more versions of this product
Product Code. 10002682
  • 3090.00 NOK / Each
Only null left
Estimated Shipment: 27-08-2025
Specifications
1Each
Test Sieve
Stainless Steel
For separation, fractioning, particle size determination
Stainless Steel
50 mm
ISO 3310/2
40mm
305 mm
Stainless Steel
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
RETSCH 305 dia. x 40 mmH Stainless Steel Test Sieve with Square Holes > Pore Size: 50 mm