Learn More
Sampling Systems™ 304 Stainless Steel Storage Containers without Toggle Clamps
These 304 Stainless Steel Storage Containers are an excellent option for multi-purpose storage in the laboratory.
Brand: Sampling Systems™ A454-100
This item is not returnable.
View return policy
Description
- Manufactured from durable 304 grade stainless steel
- Crevice-free interior
- Close fitting recessed lid
- Each with two sturdy handles
- Flat bottom with chime ring to give added protection
- Light duty construction
- Available in a wide range of capacities to fit your specific needs
Specifications
Yes | |
Recessed Lid with Handle | |
Multi-purpose Storage | |
675 mm |
Storage container | |
100 L | |
Stainless Steel | |
445 mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Sampling Systems™ 304 Stainless Steel Storage Containers without Toggle Clamps > Capacity: 10L