Learn More
Tenak™ 30 mm Cryobox Dividers
Dividers for use with 50 mm high cryoboxes.
471.00 NOK - 656.00 NOK
Specifications
Type | Grid Divider |
---|---|
For Use With (Equipment) | 50 mm high cryoboxes |
Height (Metric) | 30 mm |
Product Code | Brand | Holds | Tube Array | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Tube Array | Price | Quantity & Availability | |||||
15210464
|
Tenak™
TE22685 |
Vials with 15.50 mm O.D. | 8 x 8 cells |
600.00 NOK
Pack of 36 |
Estimated Shipment: 26-08-2025
Log in to see stock. |
|||||
15210544
|
Tenak™
TE22111 |
Vials with 41 mm O.D. | 3 x 3 cells |
471.00 NOK
Pack of 36 |
Estimated Shipment: 26-08-2025
Log in to see stock. |
|||||
15220544
|
Tenak™
TE22112 |
Vials with 31 mm O.D. | 4 x 4 cells |
471.00 NOK
Pack of 36 |
Estimated Shipment: 26-08-2025
Log in to see stock. |
|||||
15220624
|
Tenak™
TE22679 |
Vials with 7.50 mm O.D. | 16 x 16 cells |
656.00 NOK
Pack of 36 |
Estimated Shipment: 26-08-2025
Log in to see stock. |
|||||
15230544
|
Tenak™
TE22113 |
Vials with 24 mm O.D. | 5 x 5 cells |
471.00 NOK
Pack of 36 |
Estimated Shipment: 11-07-2025
Log in to see stock. |
|||||
15230624
|
Tenak™
TE22680 |
Vials with 8.75 mm O.D. | 14 x 14 cells |
656.00 NOK
Pack of 36 |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
15240624
|
Tenak™
TE22681 |
Vials with 9.50 mm O.D. | 13 x 13 cells |
656.00 NOK
Pack of 36 |
Estimated Shipment: 26-08-2025
Log in to see stock. |
|||||
15250544
|
Tenak™
TE22116 |
Vials with 9 mm O.D. | 13 x 13 cells |
600.00 NOK
Pack of 36 |
Estimated Shipment: 26-08-2025
Log in to see stock. |
|||||
15250624
|
Tenak™
TE22682 |
Vials with 10.25 mm O.D. | 12 x 12 cells |
656.00 NOK
Pack of 36 |
Estimated Shipment: 26-08-2025
Log in to see stock. |
|||||
15260544
|
Tenak™
TE22117 |
Vials with 8 mm O.D. | 14 x 14 cells |
600.00 NOK
Pack of 36 |
Estimated Shipment: 26-08-2025
Log in to see stock. |
|||||
15270544
|
Tenak™
TE22118 |
Vials with 7 mm O.D. | 16 x 16 cells |
600.00 NOK
Pack of 36 |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
15270624
|
Tenak™
TE22688 |
Vials with 25.25 mm O.D. | 5 x 5 cells |
544.00 NOK
Pack of 36 |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
15280624
|
Tenak™
TE22689 |
Vials with 31.75 mm O.D. | 4 x 4 cells |
544.00 NOK
Pack of 36 |
Estimated Shipment: 26-08-2025
Log in to see stock. |
|||||
15290624
|
Tenak™
TE22690 |
Vials with 42.50 mm O.D. | 3 x 3 cells |
544.00 NOK
Pack of 36 |
Estimated Shipment: 26-08-2025
Log in to see stock. |
|||||
11541775
|
Tenak™
TE22686 |
Vials with 17.75 mm O.D. | 7 x 7 cells |
600.00 NOK
Pack of 36 |
Estimated Shipment: 26-08-2025
Log in to see stock. |
|||||
11591675
|
Tenak™
TE22115 |
Vials with 10 mm O.D. | 12 x 12 cells |
600.00 NOK
Pack of 36 |
Estimated Shipment: 26-08-2025
Log in to see stock. |
|||||
Description
- For use in cardboard cryoboxes
- Holds cryogenic vials
- Grid style divider
Specifications
Grid Divider | |
50 mm high cryoboxes | |
30 mm |
Product Title
Tenak™ 30 mm Cryobox Dividers