missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Trifluoromethyl)benzyl alcohol, 97%
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Density | 1.293 |
Boiling Point | 79°C to 80°C (3 mmHg) |
Flash Point | 84°C (183°F) |
Refractive Index | 1.46 |
Quantity | 5 g |
Beilstein | 1450161 |
Formula Weight | 176.14 |
Percent Purity | 97% |
Chemical Name or Material | 3-(Trifluoromethyl)benzyl alcohol |
Safety and Handling

Product Identifier
- 3-(Trifluoromethyl)benzyl alcohol
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : 3-(Trifluoromethyl)benzylic alcohol
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3-(Trifluoromethyl)benzyl alcohol, 97% >
Spot an opportunity for improvement?Share a Content Correction