Learn More
3-Pyrroline, tech. 85% (remainder pyrrolidine) Product Code.: 11312019
3-Pyrroline, tech. 85% (remainder pyrrolidine)
Product Code. 11312019
5 g, 5g
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11312019
Brand: Thermo Scientific Alfa Aesar H26072.06
Request Bulk or Custom Format
CAS: 109-96-6 | C4H7N | 69.107 g/mol
3-Pyrroline is suitable to the core-level binding energies of simple unsaturated organic molecules bonded to the Si(001) surface by X-ray photoelectron spectroscopy (XPS). It is used in the synthesis of renin inhibitors and vasodilators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Pyrroline is suitable to the core-level binding energies of simple unsaturated organic molecules bonded to the Si(001) surface by X-ray photoelectron spectroscopy (XPS). It is used in the synthesis of renin inhibitors and vasodilators.
Solubility
Soluble in alcohol and ether. Miscible with water.
Notes
Air sensitive. Store at -20°C. Store away from oxidizing agents, air.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 109-96-6
Molecular Formula C4H7N
Molecular Weight (g/mol) 69.107
MDL Number MFCD00005213
InChI Key JVQIKJMSUIMUDI-UHFFFAOYSA-N
PubChem CID 66059
ChEBI CHEBI:20198
IUPAC Name 2,5-dihydro-1H-pyrrole
SMILES C1C=CCN1
Specifications
Density 0.902
Boiling Point 90°C to 91°C
Flash Point −18°C (0°F)
Odor Unpleasant
Refractive Index 1.469
Quantity 5 g
UN Number UN2733
Beilstein 103173
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,8017
Synonym 3-pyrroline, 2,5-dihydropyrrole, 1h-pyrrole, 2,5-dihydro, unii-n0kyf81sdy, delta 3-pyrroline, .delta.3-pyrroline, n0kyf81sdy, delta3-pyrroline, 2,5-dihydropyrrol, 2,5-dihydro pyrrole
Solubility Information Soluble in alcohol and ether. Miscible with water.
Formula Weight 69.11
Percent Purity 85%
Grade Technical
Assay (remainder pyrrolidine)
Chemical Name or Material 3-Pyrroline
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Product Code
Lot Number10184869 Certificate TypeCertificate of Analysis Date21/06/2023 Product CodeH26072.03,H26072.14,H26072.06,H26072
Lot Number10145391 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH26072.03,H26072.14,H26072.06
Lot Number10184869 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH26072.03,H26072.14,H26072.06
Product Title
3-Pyrroline, tech. 85% (remainder pyrrolidine) >