Learn More
3-Pyrroline, 96%, Thermo Scientific Chemicals Product Code.: 11309125
3-Pyrroline, 96%, Thermo Scientific Chemicals
Product Code. 11309125
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
Product Code. 11309125
Brand: Thermo Scientific Alfa Aesar L16349.06
Request Bulk or Custom Format
3-Pyrroline is suitable for use in a study to investigate the core-level binding energies of simple unsaturated organic molecules bonded to the Si(001) surface by X-ray photoelectron spectroscopy (XPS). It may be used in the synthesis of renin inhibitors2 and vasodilators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Pyrroline is suitable for use in a study to investigate the core-level binding energies of simple unsaturated organic molecules bonded to the Si(001) surface by X-ray photoelectron spectroscopy (XPS). It may be used in the synthesis of renin inhibitors2 and vasodilators.
Solubility
Fully misicble in water.
Notes
Air Sensitive & Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, water, moisture, acids, heat.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 109-96-6
Molecular Formula C4H7N
Molecular Weight (g/mol) 69.107
MDL Number MFCD00005213
InChI Key JVQIKJMSUIMUDI-UHFFFAOYSA-N
PubChem CID 66059
ChEBI CHEBI:20198
IUPAC Name 2,5-dihydro-1H-pyrrole
SMILES C1C=CCN1
Specifications
Density 0.902
Boiling Point 90°C to 91°C
Flash Point −18°C (0°F)
Refractive Index 1.469
Quantity 5 g
UN Number UN2733
Beilstein 103173
Sensitivity Hygroscopic
Merck Index 14,8017
Synonym 3-pyrroline, 2,5-dihydropyrrole, 1h-pyrrole, 2,5-dihydro, unii-n0kyf81sdy, delta 3-pyrroline, .delta.3-pyrroline, n0kyf81sdy, delta3-pyrroline, 2,5-dihydropyrrol, 2,5-dihydro pyrrole
Solubility Information Fully misicble in water.
Formula Weight 69.11
Percent Purity 96%
Chemical Name or Material 3-Pyrroline
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252834 Certificate TypeCertificate of Analysis Date22/10/2024 Catalog NumberL16349.03,L16349.06
Lot Number10238527 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberL16349.03,L16349.06,L16349.MD
Lot Number10146479 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL16349.03,L16349.06
Lot NumberUSLY000189 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL16349.03,L16349.06
Lot Number10144755 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL16349.03,L16349.06
Product Title
3-Pyrroline, 96%, Thermo Scientific Chemicals >