missing translation for 'onlineSavingsMsg'
Learn More
3-Phenoxybenzyl chloride, 97% Product Code.: 11417607

3-Phenoxybenzyl chloride, 97%

Product Code. 11417607
5 g, 5g
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11417607

Brand: Thermo Scientific Alfa Aesar A15670.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 53874-66-1 | C13H11ClO | 218.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53874-66-1
Molecular Formula C13H11ClO
Molecular Weight (g/mol) 218.68
MDL Number MFCD00040866
InChI Key QUYVTGFWFHQVRO-UHFFFAOYSA-N
Synonym 3-phenoxybenzyl chloride, 1-chloromethyl-3-phenoxybenzene, m-phenoxybenzyl chloride, benzene, 1-chloromethyl-3-phenoxy, 3-phenoxyphenylmethyl chloride, 3-phenoxybenzylchloride, 3-chloromethyl-1-phenoxybenzene, 1-chlormethyl-3-phenoxybenzol, acmc-1asdb, m-phenoxy-benzyl chloride
PubChem CID 93291
IUPAC Name 1-(chloromethyl)-3-phenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=CC(=C2)CCl

Specifications

Density 1.189
Boiling Point 141°C to 142°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.591
Quantity 5 g
UN Number UN3265
Beilstein 2212101
Formula Weight 218.69
Percent Purity 97%
Chemical Name or Material 3-Phenoxybenzyl chloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberUSLF005763 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeA15670.03,A15670.14,A15670.06
Lot NumberLF005763 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeA15670.03,A15670.14,A15670.06
Lot NumberI6359A Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeA15670.03,A15670.14,A15670.06
Lot NumberJ8031B Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeA15670.03,A15670.14,A15670.06
Lot Number10116710 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeA15670.03,A15670.14,A15670.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Phenoxybenzyl chloride, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.