missing translation for 'onlineSavingsMsg'
Learn More
3-Pentanol, 98% Product Code.: 10477710

3-Pentanol, 98%

Product Code. p-3758633 Shop All Thermo Scientific Chemicals Products
25 g, Glass bottle, 25g
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10477710

Brand: Thermo Scientific Acros 129990250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 584-02-1 | C5H12O | 88.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-02-1
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004574
InChI Key AQIXEPGDORPWBJ-UHFFFAOYSA-N
Synonym 3-pentanol, diethyl carbinol, sec-pentanol, pentanol-3, 3-pentyl alcohol, 3-hydroxypentane, unii-x4elc182i5, 1-ethyl-1-propanol, c2h5 2choh, isoamyl alcohol primary/secondary
PubChem CID 11428
ChEBI CHEBI:77519
IUPAC Name pentan-3-ol
SMILES CCC(CC)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -75°C
Color Colorless
Density 0.8150g/mL
Boiling Point 114°C to 116°C
Flash Point 30°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5CH(OH)C2H5
Refractive Index 1.4090 to 1.411
Quantity 25 g
Beilstein 01, 385
Merck Index 15, 7232
Specific Gravity 0.815
Solubility Information Solubility in water: 5.5g/100mL (30°C). Other solubilities: soluble in alcohol and ether
Formula Weight 88.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Pentanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0471115 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code129990250,129991000,129995000
Lot NumberA0467507 Certificate TypeCertificate of Analysis Date30/01/2025 Product Code129990250,129991000,129995000
Lot NumberA0465151 Certificate TypeCertificate of Analysis Date05/08/2024 Product Code129990250,129991000,129995000
Lot NumberA0462757 Certificate TypeCertificate of Analysis Date15/05/2024 Product Code129990250,129991000,129995000
Lot NumberA0440197 Certificate TypeCertificate of Analysis Date21/09/2023 Product Code129990250,129991000,129995000
compliance-icons compliance-icons
Product Identifier
  • 3-Pentanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Pentanol, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.