Learn More
3-Methyl(thiobenzamide), 97%, Thermo Scientific™
Product Code. 11435820
Click to view available options
Quantity:
1 g
5 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2362-63-2
Molecular Formula C8H9NS
Molecular Weight (g/mol) 151.227
MDL Number MFCD01314035
InChI Key NUFFXGAGGYWFAV-UHFFFAOYSA-N
Synonym 3-methylbenzothioamide, 3-methyl-thiobenzamide, 3-methylbenzene-1-carbothioamide, 3-methylthiobenzamide, 3-methyl thiobenzamide, 3-methylbenzenethioamide, benzenecarbothioamide, 3-methyl, amino 3-methylphenyl methane-1-thione, thio-m-toluthioamide, acmc-20amzv
PubChem CID 736826
IUPAC Name 3-methylbenzenecarbothioamide
SMILES CC1=CC=CC(=C1)C(=S)N
Specifications
Melting Point 87°C to 89°C
Quantity 5 g
Formula Weight 151.23
Percent Purity 97%
Chemical Name or Material 3-Methyl(thiobenzamide)