missing translation for 'onlineSavingsMsg'
Learn More
3-Methyl-5-hexen-3-ol, 98% Product Code.: 11414417

3-Methyl-5-hexen-3-ol, 98%

Product Code. 11414417
5 g, 5g
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11414417

Brand: Thermo Scientific Alfa Aesar B20447.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1569-44-4 | C7H14O | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1569-44-4
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00021826
InChI Key GBARKLJMQRUBKV-UHFFFAOYNA-N
Synonym 3-methyl-5-hexen-3-ol, methylaethylallylcarbinol, methylethylallylcarbinol, 5-hexen-3-ol, 3-methyl, methylaethylallylcarbinol german, 5-hexen-3-ol, 3-methyl-, 3r, acmc-20m3dr, acmc-1bx1u, 3-methyl-5-hexene-3-ol
PubChem CID 15288
IUPAC Name 3-methylhex-5-en-3-ol
SMILES CCC(C)(O)CC=C

Specifications

Density 0.84
Boiling Point 138°C to 139°C
Flash Point 44°C (111°F)
Quantity 5 g
UN Number UN1987
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material 3-Methyl-5-hexen-3-ol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Methyl-5-hexen-3-ol, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.