missing translation for 'onlineSavingsMsg'
Learn More
3-Methyl-1,5-pentanediol, 99% Product Code.: 11488917

3-Methyl-1,5-pentanediol, 99%

Product Code. p-7025182
5 g, 5g
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11488917

Brand: Thermo Scientific Alfa Aesar B20546.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4457-71-0 | C6H14O2 | 118.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4457-71-0
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.176
MDL Number MFCD00039547
InChI Key SXFJDZNJHVPHPH-UHFFFAOYSA-N
Synonym 3-methyl-1,5-pentanediol, 1,5-pentanediol, 3-methyl, 1,5-dihydroxy-3-methylpentane, unii-88495m4hns, acmc-1ai92, 3-methyl-1,5-pentane diol, 3-methyl-pentane-1,5-diol, 3-methylpentane-1,5-diol, 1,5-pentanediol,3-methyl, 4-01-00-02571 beilstein handbook referenceShow MoreShow Less
PubChem CID 20524
IUPAC Name 3-methylpentane-1,5-diol
SMILES CC(CCO)CCO

Specifications

Density 0.97
Flash Point 143°C (289°F)
Refractive Index 1.454
Quantity 5 g
Beilstein 1697331
Formula Weight 118.18
Percent Purity 99%
Chemical Name or Material 3-Methyl-1,5-pentanediol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberL01X011 Certificate TypeCertificate of Analysis Date13/01/2025 Product CodeB20546.14,B20546.06
Lot Number10245945 Certificate TypeCertificate of Analysis Date09/05/2023 Product CodeB20546.06
Lot NumberJ1430B Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeB20546.14,B20546.06
Lot NumberE4602B Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeB20546.14,B20546.06
Lot NumberL01X011 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeB20546.14,B20546.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Methyl-1,5-pentanediol, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.