missing translation for 'onlineSavingsMsg'
Learn More
3-Fluoropyridine-4-boronic acid, 98% Product Code.: 15459877

3-Fluoropyridine-4-boronic acid, 98%

Product Code. 15459877
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15459877

Brand: Thermo Scientific Alfa Aesar H53109.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 458532-97-3 | C5H5BFNO2 | 140.908 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 458532-97-3
Molecular Formula C5H5BFNO2
Molecular Weight (g/mol) 140.908
MDL Number MFCD03788558
InChI Key QUHSESYLMZVXCN-UHFFFAOYSA-N
Synonym 3-fluoropyridine-4-boronic acid, 3-fluoropyridin-4-yl boronic acid, 3-fluoropyridin-4-yl-4-boronic acid, 3-fluoro-4-pyridyl boronic acid, boronic acid, 3-fluoro-4-pyridinyl, 3-fluoro-4-pyridylboronic acid, 3-fluoropyridin-4-yl boranediol, 3-fluoro-4-pyridineboronic acid, 3-fluoropyridine-4-boronicacid
PubChem CID 2779353
IUPAC Name (3-fluoropyridin-4-yl)boronic acid
SMILES B(C1=C(C=NC=C1)F)(O)O

Specifications

Melting Point 237°C to 239°C
Quantity 250 mg
Formula Weight 140.91
Percent Purity 98%
Chemical Name or Material 3-Fluoropyridine-4-boronic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM09F050 Certificate TypeCertificate of Analysis Date18/01/2025 Catalog NumberH53109.MD,H53109.03
Lot NumberB-BI270501A Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53109.MD,H53109.03
Lot NumberM09F050 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53109.MD,H53109.03
Lot NumberU28E059 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53109.MD,H53109.03
Lot NumberZ22B043 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53109.MD,H53109.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Fluoropyridine-4-boronic acid, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.