missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-L-tyrosine, 97%, Thermo Scientific Chemicals

Product Code. 11373736
Click to view available options
Quantity:
50 mg
250 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

3-Fluoro-L-tyrosine is used as pharma and intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluoro-L-tyrosine is used as pharma and intermediates.

Solubility
Soluble in water (9.8 g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7423-96-3
Molecular Formula C9H10FNO3
Molecular Weight (g/mol) 199.18
MDL Number MFCD00002607
InChI Key VIIAUOZUUGXERI-JLDDOWRYNA-N
Synonym 3-fluoro-l-tyrosine, s-2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid, 3-fluorotyrosine, 3-fluorotyrosine, l, h-tyr 3-f-oh, unii-174nrg3m2x, s-3-fluorotyrosine, 3'-fluoro-l-tyrosine, 2s-2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid, 3-fluoro-tyrosine
PubChem CID 643330
ChEBI CHEBI:46534
IUPAC Name (2S)-2-amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C(F)=C1)C(O)=O

Specifications

Melting Point 260°C to 261°C
UN Number UN2811
Beilstein 3204802
Formula Weight 199.18
Percent Purity 97%
Quantity 250 mg
Optical Rotation −7° (c=1 in 1M HCl)
Chemical Name or Material 3-Fluoro-L-tyrosine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'