missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(trifluoromethoxy)benzoic acid, 97%, Thermo Scientific™

Product Code. 11396679
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11396679

Brand: Thermo Scientific Alfa Aesar H26234.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Fluoro-4-(trifluoromethoxy)benzoic acid is used as a reagent in the synthesis of aminoazaheterocyclic carboxamides useful in the treatment of hyper proliferative diseases.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886498-89-1
Molecular Formula C8H4F4O3
Molecular Weight (g/mol) 224.111
MDL Number MFCD06660202
InChI Key PIBFTHWKVKHHMA-UHFFFAOYSA-N
Synonym 3-fluoro-4-trifluoromethoxy benzoic acid, 3-fluoro-4-trifluoromethoxy-benzoic acid, 3-fluoro-4-trifluoromethoxy benzoicacid, benzoic acid,3-fluoro-4-trifluoromethoxy, benzoic acid, 3-fluoro-4-trifluoromethoxy
PubChem CID 2783356
IUPAC Name 3-fluoro-4-(trifluoromethoxy)benzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)F)OC(F)(F)F

Specifications

Melting Point 98°C to 101°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 224.11
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-(trifluoromethoxy)benzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.