missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methylbenzyl bromide, 97%, Thermo Scientific™

Product Code. 11441250
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145075-44-1
Molecular Formula C8H8BrF
Molecular Weight (g/mol) 203.05
MDL Number MFCD01631534
InChI Key YOACSNDETIDTAY-UHFFFAOYSA-N
Synonym 3-fluoro-4-methylbenzyl bromide, 4-bromomethyl-2-fluoro-1-methylbenzene, benzene, 4-bromomethyl-2-fluoro-1-methyl, acmc-1c02p, 3-fluoro-4-methyl-benzyl bromide, 3-fluoro-4-methylbenzylbromide, benzene,4-bromomethyl-2-fluoro-1-methyl, 4-bromomethyl-2-fluoranyl-1-methyl-benzene
PubChem CID 2774603
IUPAC Name 4-(bromomethyl)-2-fluoro-1-methylbenzene
SMILES CC1=C(F)C=C(CBr)C=C1

Specifications

Boiling Point 77°C (3 mmHg)
Refractive Index 1.546
Quantity 1 g
UN Number UN3265
Formula Weight 203.06
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-methylbenzyl bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Fluoro-4-methylbenzyl bromide, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.