missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluoro-4-methoxybenzylamine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3-Fluoro-4-methoxybenzylamine is used as an intermediate in organic synthesis.
Solubility
Immiscible with water.
Notes
Air sensitive. Store in a cool place. Incompatible with oxidizing agents, bases, acids and carbon dioxide.
3-Fluoro-4-methoxybenzylamine is used as an intermediate in organic synthesis.
Solubility
Immiscible with water.
Notes
Air sensitive. Store in a cool place. Incompatible with oxidizing agents, bases, acids and carbon dioxide.

Specifications
Specifications
Refractive Index | 1.528 |
Quantity | 1 g |
UN Number | UN2735 |
Sensitivity | Air sensitive; May form a precipitate |
Solubility Information | Immiscible with water. |
Formula Weight | 155.17 |
Percent Purity | 97% |
Chemical Name or Material | 3-Fluoro-4-methoxybenzylamine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3-Fluoro-4-methoxybenzylamine, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction