missing translation for 'onlineSavingsMsg'
Learn More
3-Fluoro-4-(2-pyridylcarbamoyl)benzeneboronic acid, 97% Product Code.: 15420796

3-Fluoro-4-(2-pyridylcarbamoyl)benzeneboronic acid, 97%

Product Code. p-7035896
250 mg, 250mg
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15420796

Brand: Thermo Scientific Alfa Aesar H54122.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1449133-22-5 | C12H10BFN2O3 | 260.031 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1449133-22-5
Molecular Formula C12H10BFN2O3
Molecular Weight (g/mol) 260.031
MDL Number MFCD20265181
InChI Key CIQKUPRZPUUGGK-UHFFFAOYSA-N
Synonym 3-fluoro-4-2-pyridylcarbamoyl benzeneboronic acid, 3-fluoro-4-pyridin-2-yl carbamoyl phenylboronic acid, 3-fluoro-4-2-pyridylcarbamoyl phenylboronic acid
PubChem CID 73995642
IUPAC Name [3-fluoro-4-(pyridin-2-ylcarbamoyl)phenyl]boronic acid
SMILES B(C1=CC(=C(C=C1)C(=O)NC2=CC=CC=N2)F)(O)O

Specifications

Quantity 250 mg
Formula Weight 260.03
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-(2-pyridylcarbamoyl)benzeneboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Fluoro-4-(2-pyridylcarbamoyl)benzeneboronic acid, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.