missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanopyridine 1-oxide, 97%

CAS: 14906-64-0 | C6H5N2O | 121.12 g/mol

666.00 NOK - 1875.00 NOK

Chemical Identifiers

CAS 14906-64-0
Molecular Formula C6H5N2O
Molecular Weight (g/mol) 121.12
MDL Number MFCD00129037
InChI Key AABSWFBNBQBPEQ-UHFFFAOYNA-N
Synonym nicotinonitrile-1-oxide, 3-cyanopyridine n-oxide, 3-cyanopyridine-n-oxide, nicotinonitrile 1-oxide, 3-cyanopyridine 1-oxide, 3-cyanopyridin-1-ium-1-olate, 3-pyridinecarbonitrile, 1-oxide, acmc-1c6hr, 3-pyridinecarbonitrile,1-oxide, woovsqcalyyudo-uhfffaoysa
PubChem CID 84686
SMILES [O-][NH++]1C=C[CH-]C(=C1)C#N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11418837
SDS View Documents
Thermo Scientific Alfa Aesar
A19961.88
2.5 g
666.00 NOK
2.50g
Please to purchase this item. Need a web account? Register with us today!
11428837
SDS View Documents
Thermo Scientific Alfa Aesar
A19961.09
10 g
1875.00 NOK
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'einecsNumber' : 238-975-1

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Cyanopyridine 1-oxide, 97%

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.