Learn More
3-Chloroperoxybenzoic Acid, 70-75%, balance 3-Chlorobenzoic Acid and water
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Chemical Name or Material | 3-Chloroperoxybenzoic Acid |
Name Note | 70 - 75% |
CAS | 937-14-4, 535-80-8, 7732-18-5 |
CAS Min % | 70.0, 25.0, 25.0 |
CAS Max % | 75.0, 30.0, 30.0 |
Melting Point | 92.0°C to 94.0°C |
Color | White |
Density | 0.5600g/mL |
Assay Percent Range | di-m-Chlorobenzoyl peroxide: < 0.1% Typical (HPLC) |
Packaging | Plastic Bottle |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2625801301 | Certificate of Analysis | 29/05/2025 | 255795000,255791000,255790010,255790250 |
2624834801 | Certificate of Analysis | 29/05/2025 | 255795000,255791000,255790010,255790250 |
2624834602 | Certificate of Analysis | 29/05/2025 | 255795000,255791000,255790010,255790250 |
2624834601 | Certificate of Analysis | 29/05/2025 | 255795000,255791000,255790010,255790250 |
2624834501 | Certificate of Analysis | 29/05/2025 | 255795000,255791000,255790010,255790250 |
Safety and Handling
- 3-Chloroperoxybenzoic acid
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Organic peroxide Type C & D
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H242-Heating may cause a fire.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.