missing translation for 'onlineSavingsMsg'
Learn More
3-(Chloromethyl)pyridine hydrochloride, 97% Product Code.: 11470833

3-(Chloromethyl)pyridine hydrochloride, 97%

Product Code. p-7021085
5 g, 5g
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11470833

Brand: Thermo Scientific Alfa Aesar A13279.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6959-48-4 | C6H7Cl2N | 164.029 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6959-48-4
Molecular Formula C6H7Cl2N
Molecular Weight (g/mol) 164.029
MDL Number MFCD00012818
InChI Key UZGLOGCJCWBBIV-UHFFFAOYSA-N
Synonym 3-chloromethyl pyridine hydrochloride, 3-chloromethylpyridine hydrochloride, 3-picolyl chloride hydrochloride, 3-chloromethyl pyridine hcl, ccris 140, pyridine, 3-chloromethyl-, hydrochloride, 3-pyridylmethyl chloride hydrochloride, unii-bqa8fyp4yn, 3-chloromethyl pyridinium chloride, 3-chloromethyl pyridine.hcl
PubChem CID 23394
IUPAC Name 3-(chloromethyl)pyridine;hydrochloride
SMILES C1=CC(=CN=C1)CCl.Cl

Specifications

Melting Point 136°C to 142°C
pH 3
Quantity 5 g
UN Number UN3261
Beilstein 3689156
Sensitivity Hygroscopic
Formula Weight 164.04
Percent Purity 97%
Chemical Name or Material 3-(Chloromethyl)pyridine hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number10099924 Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA13279.22,A13279.06,A13279.14
Lot NumberG2960B Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA13279.22,A13279.06,A13279.14
Lot Number90006324 Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA13279.22,A13279.06,A13279.14
Lot Number10227450 Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA13279.22,A13279.06,A13279.14
Lot Number10026228 Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA13279.22,A13279.06,A13279.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-(Chloromethyl)pyridine hydrochloride, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.