missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chlorobenzotrifluoride, 98%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3-Chlorobenzotrifluoride is used as an intermediate in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water <0.1 g/100 mL at 22°C.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
3-Chlorobenzotrifluoride is used as an intermediate in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water <0.1 g/100 mL at 22°C.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.

Specifications
Specifications
Melting Point | -56°C |
Boiling Point | 137°C to 138°C |
Flash Point | 38°C (100°F) |
Quantity | 25 g |
UN Number | UN2234 |
Beilstein | 510215 |
Formula Weight | 180.56 |
Percent Purity | 98% |
Chemical Name or Material | 3-Chlorobenzotrifluoride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3-Chlorobenzotrifluoride, 98%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction