missing translation for 'onlineSavingsMsg'
Learn More
3-Chloro-4-methylphenylhydrazine hydrochloride, 97%, Thermo Scientific™ Product Code.: 11459547

3-Chloro-4-methylphenylhydrazine hydrochloride, 97%, Thermo Scientific™

Product Code. 11459547
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11459547

Brand: Thermo Scientific Alfa Aesar B22053.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54812-56-5
Molecular Formula C7H10Cl2N2
Molecular Weight (g/mol) 193.071
MDL Number MFCD00012936
InChI Key DQDFVQJOXLGGBL-UHFFFAOYSA-N
Synonym 3-chloro-4-methylphenylhydrazine hydrochloride, 3-chloro-p-tolylhydrazine hydrochloride, 3-chloro-4-methylphenyl hydrazine hydrochloride, 3-chloro-4-methylphenyl hydrazine monohydrochloride, hydrazine, 3-chloro-4-methylphenyl-, monohydrochloride, hydrazine, 3-chloro-4-methylphenyl-, hydrochloride, 3-chloro-4-methylphenyl hydrazine hcl, acmc-1alsi, ksc252c4h, 3-chloro-4-methylphenylhydrazine hcl
PubChem CID 3085075
IUPAC Name (3-chloro-4-methylphenyl)hydrazine;hydrochloride
SMILES CC1=C(C=C(C=C1)NN)Cl.Cl

Specifications

Melting Point ∼233°C (decomposition)
Quantity 1 g
Formula Weight 193.08
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-methylphenylhydrazine hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Chloro-4-methylphenylhydrazine hydrochloride, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.