Learn More
Thermo Scientific Alfa Aesar 1GR 3-Chloro-4-(cyclohexylcarbamoyl)benzeneboronic acid, 97% 1g
Brand: Thermo Scientific Alfa Aesar H54149.03
1GR 3-Chloro-4-(cyclohexylcarbamoyl)benzeneboronic acid, 97% 1g
Additional Details : CAS Number : 957034-65-0 Weight : 0.00100kg
Description

Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
B-BF220602 | Certificate of Analysis | 07/12/2021 |