missing translation for 'onlineSavingsMsg'
Learn More
3-Buten-1-ol, 98+% Product Code.: 10522841

3-Buten-1-ol, 98+%

Product Code. p-3735788 Shop All Thermo Scientific Chemicals Products
25 mL, Glass bottle, 25mL
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
This item is not returnable. View return policy

Product Code. 10522841

Brand: Thermo Scientific Acros 295750250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 627-27-0 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-27-0
MDL Number MFCD00002959
InChI Key ZSPTYLOMNJNZNG-UHFFFAOYSA-N
Synonym 3-buten-1-ol, allylcarbinol, 3-butenyl alcohol, 1-buten-4-ol, vinylethyl alcohol, 3-butene-1-ol, ch2=chch2ch2oh, homoallyl alcohol, unii-3db2krm1i9, 1-hydroxy-3-butene
PubChem CID 69389
IUPAC Name but-3-en-1-ol
SMILES C=CCCO

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8300g/mL
Boiling Point 112.0°C to 114.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Molecular Formula C4H8O
Linear Formula H2C=CHCH2CH2OH
Refractive Index 1.4200 to 1.4220
Quantity 25 mL
Beilstein 01, 441
Specific Gravity 0.83
Solubility Information Solubility in water: soluble in water. Other solubilities: miscible with ethanol,ether,acetone
Molecular Weight (g/mol) 72.11
Formula Weight 72.11
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 3-Buten-1-ol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0460713 Certificate TypeCertificate of Analysis Date28/02/2024 Product Code295751000,295755000,295750250
Lot NumberA0443294 Certificate TypeCertificate of Analysis Date22/12/2023 Product Code295751000,295755000,295750250
Lot NumberA0422626 Certificate TypeCertificate of Analysis Date31/03/2022 Product Code295751000,295755000,295750250
Lot NumberA0399399 Certificate TypeCertificate of Analysis Date10/08/2021 Product Code295751000,295755000,295750250
compliance-icons compliance-icons
Product Identifier
  • 3-Buten-1-ol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Buten-1-ol, 98+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.