missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-(trifluoromethoxy)benzyl bromide, 97%, Thermo Scientific™

Product Code. 15444947
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1031929-10-8
Molecular Formula C8H5Br2F3O
Molecular Weight (g/mol) 333.93
MDL Number MFCD11035864
InChI Key QYRNFJCHQHPVTI-UHFFFAOYSA-N
Synonym 3-bromo-5-trifluoromethoxy benzyl bromide, 1-bromo-3-bromomethyl-5-trifluoromethoxy benzene
PubChem CID 26985713
IUPAC Name 1-bromo-3-(bromomethyl)-5-(trifluoromethoxy)benzene
SMILES C1=C(C=C(C=C1OC(F)(F)F)Br)CBr

Specifications

Refractive Index 1.5185
Quantity 1 g
UN Number UN3265
Formula Weight 333.93
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-(trifluoromethoxy)benzyl bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'