missing translation for 'onlineSavingsMsg'
Learn More
3-Bromo-4-(trifluoromethoxy)aniline, 97%, Thermo Scientific™ Product Code.: 11356559

3-Bromo-4-(trifluoromethoxy)aniline, 97%, Thermo Scientific™

Product Code. 11356559
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11356559

Brand: Thermo Scientific Alfa Aesar H26174.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 191602-54-7
Molecular Formula C7H5BrF3NO
Molecular Weight (g/mol) 256.022
MDL Number MFCD00190127
InChI Key RAQMUBDHNKQNTD-UHFFFAOYSA-N
Synonym 3-bromo-4-trifluoromethoxy aniline, 3-bromo-4-trifluoromethoxy benzenamine, 4-amino-2-bromo trifluoromethoxy benzene, 3-bromo-4-trifluoromethoxy-phenylamine, 2-bromo-4-amino-trifluoromethoxybenzene, benzenamine, 3-bromo-4-trifluoromethoxy, 1-amino-3-bromo-4-trifluoromethoxy benzene, pubchem2925, ksc496c0p
PubChem CID 2736418
IUPAC Name 3-bromo-4-(trifluoromethoxy)aniline
SMILES C1=CC(=C(C=C1N)Br)OC(F)(F)F

Specifications

Quantity 1 g
UN Number UN2810
Formula Weight 256.02
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-(trifluoromethoxy)aniline

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Product Code
Lot NumberD11T004 Certificate TypeCertificate of Analysis Date07/12/2021 Product Code
Lot Number10164248 Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
Lot Number10144914 Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
Lot Number10157883 Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Bromo-4-(trifluoromethoxy)aniline, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.