missing translation for 'onlineSavingsMsg'
Learn More
3-(Boc-amino)-3-(3-fluorophenyl)propionic acid, 99%, Thermo Scientific™ Product Code.: 11389775

3-(Boc-amino)-3-(3-fluorophenyl)propionic acid, 99%, Thermo Scientific™

Product Code. p-7047823
250 mg, 250mg
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11389775

Brand: Thermo Scientific Alfa Aesar L19805.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 284493-59-0
Molecular Formula C14H18FNO4
Molecular Weight (g/mol) 283.299
MDL Number MFCD02090701
InChI Key IQPQPXUDXQDVMK-UHFFFAOYSA-N
Synonym 3-tert-butoxycarbonyl amino-3-3-fluorophenyl propanoic acid, 3-boc-amino-3-3-fluorophenyl propionic acid, 3-n-boc-amino-3-3-fluorophenyl propionic acid, 3-3-fluorophenyl-3-2-methylpropan-2-yl oxycarbonylamino propanoic acid, boc-3-amino-3-3-fluorophenyl propionic acid, 3-tert-butoxy carbonylamino-3-3-fluorophenyl propanoic acid, acmc-20anb3, boc-r-3-amino-3-3-fluoro-phenyl-propionic acid, r,s-3-amino-3-3-fluorophenyl propanoic acid
PubChem CID 2783065
IUPAC Name 3-(3-fluorophenyl)-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC(=CC=C1)F

Specifications

Melting Point 124°C to 126°C
Quantity 250 mg
Formula Weight 283.3
Percent Purity 99%
Chemical Name or Material 3-(Boc-amino)-3-(3-fluorophenyl)propionic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-(Boc-amino)-3-(3-fluorophenyl)propionic acid, 99%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.