missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-cyclopropyl-1H-pyrazole, 96%, Thermo Scientific™

Product Code. 11419610
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175137-46-9
Molecular Formula C6H9N3
Molecular Weight (g/mol) 123.159
MDL Number MFCD08461681
InChI Key MXVAGCQKBDMKPG-UHFFFAOYSA-N
Synonym 3-cyclopropyl-1h-pyrazol-5-amine, 3-amino-5-cyclopropyl-1h-pyrazole, 5-cyclopropyl-2h-pyrazol-3-ylamine, 3-amino-5-cyclopropylpyrazole, 1h-pyrazol-3-amine, 5-cyclopropyl, 3-cyclopropylpyrazol-5-amine, 5-cyclopropyl-2h-pyrazol-3-amine, 3-cyclopropyl-1h-pyrazole-5-amine, 5-cyclopropyl-1h-pyrazole-3-amine, 3-cyclopropylpyrazole-5-ylamine
PubChem CID 2758014
IUPAC Name 5-cyclopropyl-1H-pyrazol-3-amine
SMILES C1CC1C2=CC(=NN2)N

Specifications

Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 123.16
Percent Purity 96%
Chemical Name or Material 3-Amino-5-cyclopropyl-1H-pyrazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Amino-5-cyclopropyl-1H-pyrazole, 96%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.