missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-iodopyridine, 98%, Thermo Scientific Chemicals

Product Code. 11454390
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105752-11-2
Molecular Formula C5H5IN2
Molecular Weight (g/mol) 220.01
MDL Number MFCD04971334
InChI Key ZJRSKTXMSIVNAU-UHFFFAOYSA-N
Synonym 3-amino-4-iodopyridine, 4-iodo-pyridin-3-ylamine, 3-pyridinamine, 4-iodo, 4-iodo-3-pyridinamine, 3-amine-4-iodopyridine, pubchem6681, 4-iodo-3-pyridylamine, 4-iodo-3-aminopyridine, 4-iodo-3-pyridinylamine, 4-iodopyridin-3-ylamine
PubChem CID 819132
IUPAC Name 4-iodopyridin-3-amine
SMILES NC1=CN=CC=C1I

Specifications

Melting Point 55°C to 68°C
Quantity 1 g
Sensitivity Air and light sensitive
Formula Weight 220.01
Percent Purity 98%
Chemical Name or Material 3-Amino-4-iodopyridine
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'